Anti-SAFB2 Rabbit Polyclonal Antibody

Supplier: Proteintech
11642-1-AP-150UL
10094-116EA 402 USD
10094-116
Anti-SAFB2 Rabbit Polyclonal Antibody
Antibodies
SAFB2, also named as Scaffold attachment factor B2 or KIAA0138, is a 953 amino acid protein, which contains one SAP domain and one RRM (RNA recognition motif) domain. SAFB2 expressed at high levels in the CNS and at low levels in the liver. It also is expressed in a wide number of breast cancer cell lines. SAFB2 binds to scaffold/matrix attachment region (S/MAR) DNA and can function as an estrogen receptor corepressor and can also inhibit cell proliferation.

Western Blot: Human Brain Tissue, 1:1000-1:10000; IHC: Human breast cancer Tissue, 1:20-1:200

Type: Primary
Antigen: SAFB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR