Anti-SEPT5 Rabbit Polyclonal Antibody

Supplier: Proteintech
11631-1-AP-150UL
10094-320EA 402 USD
10094-320
Anti-SEPT5 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Brain Tissue, 1:200-1:2000; IHC: Human gliomas Tissue, 1:20-1:200; IP: Mouse Brain Tissue, 1:200-1:1000

Type: Primary
Antigen: SEPT5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR