Anti-ERR1F1 Rabbit Polyclonal Antibody

Supplier: Proteintech
11630-1-AP-150UL
10090-226EA 599.76 USD
10090-226
Anti-ERR1F1 Rabbit Polyclonal Antibody
Antibodies
The mitogen inducible gene 6 (MIG6) is located in human chromosome 1p36, a locus frequently associated with human lung cancer. MIG6 gene expression can be rapidly induced by factors including hormones, growth factor, and stresses. MIG6 encodes ERBB receptor feedback inhibitor 1 (ERRFI1) levels of which are very low in quiescent cells but upregulated by mitogens. MIG6 may play important roles in regulating stress response, maintaining homeostasis in tissues like joints or cardiac muscle, and functioning as a tumor suppressor.

Western Blot: HeLa Cells, 1:500-1:5000; IHC: Human breast cancer Tissue, 1:20-1:200

Type: Primary
Antigen: ERR1F1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR