Anti-UHMK1 Rabbit Polyclonal Antibody

Supplier: Proteintech
11624-1-AP-150UL
10096-764EA 459.7 USD
10096-764
Anti-UHMK1 Rabbit Polyclonal Antibody
Antibodies
UHMK1, also named as KIS, or KIST, is a 419 amino acid protein, which contains one RRM (RNA recognition motif) domain, one protein kinase domain, and belongs to the protein kinase superfamily. UHMK1 is widely expressed, with highest levels in skeletal muscle, kidney, placenta and peripheral blood leukocytes. UHMK1 localizes in the nucleus and upon serum stimulation, phosphorylating CDKN1B/p27Kip1, thus controlling CDKN1B subcellular location and cell cycle progression in G1 phase. UHMK1 may be involved in trafficking and/or processing of RNA and may have a role in the developing nervous system and the adult brain.

Western Blot: Mouse Thymus Tissue, 1:500-1:5000; IHC: Human Prostate Cancer Tissue, 1:20-1:200

Type: Primary
Antigen: UHMK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR