Anti-BCL11A Rabbit Polyclonal Antibody

Supplier: Proteintech
11613-1-AP-150UL
10083-546EA 402 USD
10083-546
Anti-BCL11A Rabbit Polyclonal Antibody
Antibodies
B-CELL CLL/LYMPHOMA 11A(BCL11A)is a highly conserved zinc finger gene. It acts as a myeloid and B-cell proto-oncogene, and may play key roles in leukemogenesis and hematopoiesis. It is an essential factor in lymphopoiesis, and required for B-cell formation in fetal liver. May function as a modulator of the transcriptional repression activity of ARP1. It is also involved in lymphoid malignancies through translocation or amplification events. It has recently been shown that BCL11A expression is inversely correlated with γ globin expression.. This is a rabbit polyclonal antibody raised against part chain of N-terminal BCL11A of human origin. There are three isoforms of BCL11A: isoform1(773aa), isoform2(486aa) and isoform3(239aa)

IHC: Human gliomas, 1:20-1:200

Type: Primary
Antigen: BCL11A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR