Anti-CPSF3 Rabbit Polyclonal Antibody

Supplier: Proteintech
11609-1-AP-150UL
10085-136EA 402 USD
10085-136
Anti-CPSF3 Rabbit Polyclonal Antibody
Antibodies
Western Blot: HEK-293 Cells, 1:500-1:5000

Type: Primary
Antigen: CPSF3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR