Anti-ALDH4A1 Rabbit Polyclonal Antibody

Supplier: Proteintech
11604-1-AP-150UL
10082-766EA 402 USD
10082-766
Anti-ALDH4A1 Rabbit Polyclonal Antibody
Antibodies
ALDH4A1(Aldehyde dehydrogenase family 4 member A1) is also named as ALDH4, P5CDH,P5C dehydrogenase and belongs to the aldehyde dehydrogenase family. P5CDH is a mitochondrial matrix NAD(+)-dependent dehydrogenase which catalyzes the second step of the proline degradation pathway, converting pyrroline-5-carboxylate to glutamate.Defects in ALDH4A1 are the cause of hyperprolinemia type 2 (HP-2).

Western Blot: Human Heart Tissue, 1:500-1:5000; IHC: Human Liver Tissue, 1:20-1:200

Type: Primary
Antigen: ALDH4A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR