Anti-RGS5 Rabbit Polyclonal Antibody

Supplier: Proteintech
11590-1-AP-150UL
10093-630EA 599.76 USD
10093-630
Anti-RGS5 Rabbit Polyclonal Antibody
Antibodies
RGS5 inhibits signal transduction by increasing the GTPase activity of G protein alpha subunits thereby driving them into their inactive GDP-bound form. It binds to G(i)-alpha and G(o)-alpha, but not to G(s)-alpha. The 42kd band recognized by this antibody is a RGS5-RGS5 dimer. (Pubmed:17762159)

Western Blot: Human Heart Tissue, 1:500-1:5000; IHC: Human prostate cancer, 1:20-1:200

Type: Primary
Antigen: RGS5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR