Anti-RABL2B Rabbit Polyclonal Antibody

Supplier: Proteintech
11588-1-AP-150UL
10093-256EA 402 USD
10093-256
Anti-RABL2B Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Brain Tissue, 1:1000-1:10000; IHC: Human lung cancer, 1:20-1:200

Type: Primary
Antigen: RABL2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR