Anti-SRP68 Rabbit Polyclonal Antibody

Supplier: Proteintech
11585-1-AP-150UL
10095-176EA 459.7 USD
10095-176
Anti-SRP68 Rabbit Polyclonal Antibody
Antibodies
The signal recognition particle (SRP) is a ribonucleoprotein complex that mediates the targeting of secretory proteins to the rough endoplasmic reticulum (ER) membrane. This ribonucleoprotein complex might interact directly with the docking protein in the ER membrane and possibly participate in the elongation arrest function. SRP68 is one component of the ribonucleprotein complex, and forms a heterodimer with SRP72.

Western Blot: Human Kidney Tissue, 1:500-1:5000; IHC: Human gliomas, 1:20-1:200

Type: Primary
Antigen: SRP68
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR