Anti-OGT Rabbit Polyclonal Antibody

Supplier: Proteintech
11576-2-AP-150UL
10091-566EA 614.88 USD
10091-566
Anti-OGT Rabbit Polyclonal Antibody
Antibodies
O-linked N-acetylglucosamine transferase (OGT) catalyzes the attachment of N-acetylglucosamine (GlcNAc) monosaccharides to the hydroxyl group of serine or threonine residues of numerous nuclear and cytoplasmic proteins and may play important roles in a large number of diverse intracellular processes ranging from translational control, transcription, transcriptional repression, insulin resistance and regulation of the cell cycle. It exists as a heterotrimeric complex with two 110 kDa and one 70 kDa subunits. Recent studies have shown that O-GlcNAcylation plays essential roles in cancer formation and progression. O-GlcNAcylation as well as OGT expression was found to be significantly elevated in the cancer tissues.

Western Blot: Human Brain Tissue, 1:500-1:5000; IHC: Human pancreas cancer Tissue, 1:20-1:200; IP: Mouse Brain Tissue, 1:500-1:5000

Type: Primary
Antigen: OGT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR