Ubiquitin is an important cellular signal that targets proteins for degradation or regulates their functions. UBL7 (ORF name SB132) encodes ubiquitin-like protein7 also named bone marrow stromal cell ubiquitin-like protein (BMSC-UbP) capable of binding ubiquitin. BMSC-UbP protein is derived from bone marrow stromal cells containing a ubiquitin-associated (UBA) domain at the C terminus that has been implicated in linking cellular processes and the ubiquitin system. BMSC-UbP mRNA is widely expressed in human multiple tissues and various tumor cell lines and is decreased in BMSC stimulated with phorbol myristate acetate (PMA) suggesting a role in regulation of BMSC function.
Western Blot: HepG2 Cells, 1:500-1:5000; IHC: Human gliomas Tissue, 1:20-1:200; IP: HeLa Cells, 1:500-1:5000; IF: HepG2 Cells, 1:10-1:100
Type: Primary
Antigen: BMSC UbP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat