Anti-ING2 Rabbit Polyclonal Antibody

Supplier: Proteintech
11560-1-AP-150UL
10088-886EA 402 USD
10088-886
Anti-ING2 Rabbit Polyclonal Antibody
Antibodies
ING2, also named as ING1L, belongs to the ING family and contains a PHD-type zinc finger. It is induced by the DNA-damaging agents etoposide and neocarzinostatin. As a candidate tumor suppressor, ING2 seems to be involved in p53/TP53 activation and p53/TP53-dependent apoptotic pathways, probably by enhancing acetylation of p53/TP53. It is a component of a mSin3A-like corepressor complex, which is probably involved in deacetylation of nucleosomal histones. Recent finding reported that ING2 regulates muscle differentiation by regulating chormatin remodeling program. Catalog#11560-1-AP is a rabbit polyclonal antibody raised against full length ING2 of human origin.

Western Blot: HEK-293 Cells, 1:200-1:2000; IHC: Human Colon cancer, 1:20-1:200

Type: Primary
Antigen: ING2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR