Anti-Antizyme Inhibitor 1 Rabbit Polyclonal Antibody

Supplier: Proteintech
11548-1-AP-150UL
10083-432EA 459.7 USD
10083-432
Anti-Antizyme Inhibitor 1 Rabbit Polyclonal Antibody
Antibodies
AZIN1(Antizyme inhibitor 1) is also named as OAZI, OAZIN and belongs to the Orn/Lys/Arg decarboxylase class-II family. It catalyzes the conversion of ornithine to putrescine in the first and apparently rate-limiting step in polyamine biosynthesis, and playing a role in the regulation of polyamine synthesis (negative regulator of cellular polyamines).

Western Blot: Human Liver Tissue, 1:500-1:5000; IHC: Human gliomas Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: [delete]
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR