Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NMT1 Rabbit Polyclonal Antibody

Supplier: Proteintech
11546-1-AP
NMT1 is a N-myristoyltransferase responsible for the transfer of myristate from CoA to an amino-terminal glycine of many eukaryotic proteins, which facilitates the targeting of proteins to membrane surfaces and is essential for viability of the organism. Insertional mutagenesis of the Nmt1 gene in Saccharomyces cerevisiae causes recessive lethality. Humans and mice possess two distinct but structurally similar enzymes, NMT1 and NMT2, ubiquitously expressed in most human and mouse tissues. Western analysis revealed that there are 4 isoforms of NMT1 with apparent molecular masses ranging from 49 to 68 kD. In cell fractionation studies, the 68-kD NMT1 isoform and NMT2 were present in both membrane and cytoplasmic fractions, while the smaller NMT1 isoforms were predominantly cytoplasmic.

Western Blot: Mouse pancreas Tissue, 1:200-1:2000; IHC: Human gliomas Tissue, 1:20-1:200; IF: MCF-7 Cells, 1:20-1:200

Type: Primary
Antigen: NMT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
150 µL 11546-1-AP 10091-168 Each (150µl) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Shipping duration cannot exceed 2 days.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol NMT1
Antigen Name N-myristoyltransferase 1
Reactivity Rat, Mouse, Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Isotype IgG
Western Blot Yes
ELISA Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Immunogen Recombinant Protein
Purification Antigen affinity Purification
Storage Buffer PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
Shipping Temperature Store at -20°C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty