Anti-PLAGL2 Rabbit Polyclonal Antibody

Supplier: Proteintech
11540-1-AP-150UL
10092-426EA 402 USD
10092-426
Anti-PLAGL2 Rabbit Polyclonal Antibody
Antibodies
PLAGL2, also named as KIAA0198, belongs to the krueppel C2H2-type zinc-finger protein family. It shows weak transcriptional activatory activity. PLAGL2), a proto-oncogene, is implicated in a variety of cancers including acute myeloid leukemia (AML), malignant glioma, colon cancer, and lung adenocarcinoma.PLAGL2 can function as a tumor suppressor by initiating cell cycle arrest and apoptosis.

Western Blot: Raji Cells, 1:500-1:5000

Type: Primary
Antigen: PLAGL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR