Anti-TUSC2 Rabbit Polyclonal Antibody

Supplier: Proteintech
11538-1-AP-150UL
10096-510EA 402 USD
10096-510
Anti-TUSC2 Rabbit Polyclonal Antibody
Antibodies
FUS1 (or TUSC2) gene is a highly conserved lung cancer candidate gene, which was identified in the 120 kb 3p21.3 critical region contained in nested lung and breast cancer homozygous deletions. Overexpression of FUS1 gene leads to G1 arrest and growth inhibition of lung cancer cells . The encoded Fusion-1 protein was down-regulated, mutated or lost in the majority of inflammatory thoracic malignancies. It has been evidenced that Fusion-1 establishes its immune- and tumour-suppressive activities via regulation of mitochondrial homeostasis . In addition, myristoylation is found to be required for Fusion-1-mediated tumor-suppressing activity and suggest a novel mechanism for the inactivation of tumor suppressors in lung cancer and a role for deficient posttranslational modification in tumor suppressor-gene-mediated carcinogenesis .

Western Blot: Human Heart Tissue, 1:500-1:5000; IHC: Human lung cancer Tissue, 1:20-1:200

Type: Primary
Antigen: TUSC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR