Anti-LRRC8D Rabbit Polyclonal Antibody

Supplier: Proteintech
11537-1-AP-150UL
10089-808EA 402 USD
10089-808
Anti-LRRC8D Rabbit Polyclonal Antibody
Antibodies
LRRC8D (or LRRC5L) gene encodes a 98-kDa multi-pass membrane protein leucine-rich repeat-containing protein 8D, which contains 13 LRRs (leucine-rich repeats). The molecular function of this protein is so far not well understood. Recently, LRRC8D gene was found to be up-regulated after IL-2 deprivation in immortalised Interleukin-2(IL-2)-dependent human T cells, indicating a possible involvement of LRRC8D in cancer development. This LRRC8D antibody can’t cross-recognize LRRC8A.

Western Blot: HeLa Cells, 1:200-1:2000

Type: Primary
Antigen: LRRC8D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR