Anti-CNN3 Rabbit Polyclonal Antibody

Supplier: Proteintech
11509-1-AP-150UL
10084-928EA 402 USD
10084-928
Anti-CNN3 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Kidney Tissue, 1:200-1:2000; IHC: Human Prostate Cancer Tissue, 1:20-1:200; IF: Hela Cells, 1:10-1:100

Type: Primary
Antigen: CNN3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR