Anti-ASGR2 Rabbit Polyclonal Antibody

Supplier: Proteintech
11501-2-AP-150UL
10082-318EA 402 USD
10082-318
Anti-ASGR2 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Liver Tissue, 1:200-1:1000

Type: Primary
Antigen: ASGR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR