Anti-APLN Rabbit Polyclonal Antibody

Supplier: Proteintech
11497-1-AP-150UL
10082-990EA 402 USD
10082-990
Anti-APLN Rabbit Polyclonal Antibody
Antibodies
Apelin, isolated from bovine stomach tissue extractsis, is recognized as the endogenous ligand of the angiotensin-like-receptor 1 (APJ), and the human orphan G-protein-coupled receptor. Apelin is widely expressed in various organs such as the heart, lung, kidney, liver, adipose tissue, gastrointestinal tract, brain, adrenal glands, endothelium, and human plasma. Apelin has been found to be a potent stimulator of cardiac contractility and may function in the regulation of the cardiovascular system. Apelin is also known to be involved in the maintenance of insulin sensitivity and play an important role in liver disease.

IHC: Human pancreas cancer Tissue, 1:20-1:200

Type: Primary
Antigen: APLN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR