Anti-PTPMT1 Rabbit Polyclonal Antibody

Supplier: Proteintech
11493-1-AP-150UL
10093-054EA 402 USD
10093-054
Anti-PTPMT1 Rabbit Polyclonal Antibody
Antibodies
PTPMT1, also named as MOSP and PLIP, is a Lipid phosphatase which dephosphorylates phosphatidylglycerophosphate (PGP) to phosphatidylglycerol (PG). PTPMT1 has also been shown to display phosphatase activity toward phosphoprotein substrates, specifically mediates dephosphorylation of mitochondrial proteins, thereby playing an essential role in ATP production. Has probably a preference for proteins phosphorylated on Ser and/or Thr residues compared to proteins phosphorylated on Tyr residues. It is a mitochondrial Pten-like phosphatidylinositol phosphate (PIP) phosphatase, resulted in developmental arrest and postimplantation lethality.

Western Blot: Mouse pancreas Tissue, 1:200-1:1000; IHC: Human pancreas cancer, 1:20-1:200

Type: Primary
Antigen: PTPMT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR