Anti-PPFIA3 Rabbit Polyclonal Antibody

Supplier: Proteintech
11492-1-AP-150UL
10092-640EA 402 USD
10092-640
Anti-PPFIA3 Rabbit Polyclonal Antibody
Antibodies
PPFIA3, also named as KIAA0654, belongs to the liprin family and Liprin-alpha subfamily. PPFIA3 may regulate the disassembly of focal adhesions. It may localize receptor-like tyrosine phosphatases type 2A at specific sites on the plasma membrane, possibly regulating their interaction with the extracellular environment and their association with substrates. PPFIA3 exhibites more than 5 fold accumulations in the monoribosome fraction. PPFIA3 is decreased at the protein level by the miR-99 family but not in the luciferase reporter with the 3’UTR of PPFIA3.

Western Blot: Mouse Brain Tissue, 1:200-1:2000

Type: Primary
Antigen: PPFIA3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR