Anti-PHACS Rabbit Polyclonal Antibody

Supplier: Proteintech
11491-1-AP-150UL
10092-172EA 402 USD
10092-172
Anti-PHACS Rabbit Polyclonal Antibody
Antibodies
ACCS(1-aminocyclopropane-1-carboxylate synthase-like protein 1) is also named as PHACS and belongs to the class-I pyridoxal-phosphate-dependent aminotransferase family. The deduced 501-amino acid protein has a calculated molecular mass of 58 kD. It contains overlapping aminotransferase I and beta-eliminating lyase domains, and it shares structural similarity with aspartate aminotransferase, tyrosine aminotransferase, and enzymes that catalyze beta-elimination reactions on amino acids.

Western Blot: MDA-MB-453s Cells, 1:500-1:5000; IHC: Human Colon cancer Tissue, 1:20-1:200

Type: Primary
Antigen: PHACS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR