Anti-HECTD3 Rabbit Polyclonal Antibody

Supplier: Proteintech
11487-1-AP-150UL
10088-148EA 524.48 USD
10088-148
Anti-HECTD3 Rabbit Polyclonal Antibody
Antibodies
HECTD3(HECT domain-containing protein 3) may be involved in the regulation of mitotic metaphase/anaphase transition. It can specifically ubiquitinate TRIOBP and target it for proteasomal degradation and it can also mediates the ubiquitination of STX8. HECTD3 is a new oncogenic E3 ligase in human prostate cancer.( Li Y, etc. 2008) HECTD3 may facilitate cell cycle progression via regulating ubiquitination and degradation of Tara. It has 2 isoforms produced by alternative splicing.

Western Blot: MCF7 Cells, 1:500-1:5000

Type: Primary
Antigen: HECTD3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR