Anti-STAC Rabbit Polyclonal Antibody

Supplier: Proteintech
11480-1-AP-150UL
10095-230EA 402 USD
10095-230
Anti-STAC Rabbit Polyclonal Antibody
Antibodies
Western Blot:PC-3 Cells, 1:1000-1:10000

Type: Primary
Antigen: STAC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR