Anti-MTAP Rabbit Polyclonal Antibody

Supplier: Proteintech
11475-1-AP-150UL
10090-532EA 524.48 USD
10090-532
Anti-MTAP Rabbit Polyclonal Antibody
Antibodies
IHC: Human Colon cancer Tissue, 1:20-1:200; IP: MCF-7 Cells, 1:200-1:2000; Western Blot: MCF7 Cells, 1:200-1:2000

Type: Primary
Antigen: MTAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR