Anti-FXYD7 Rabbit Polyclonal Antibody

Supplier: Proteintech
11465-1-AP-150UL
10087-204EA 402 USD
10087-204
Anti-FXYD7 Rabbit Polyclonal Antibody
Antibodies
FXYD7 (FXYD domain containing ion transport regulator 7) is a single-pass membrane protein that belongs to the FXYD family. The FXYD family, which contains seven members, are tissue specific regulators of the Na,K-ATPase. Expressed exclusively in the brain, FXYD7 is an isoform-specific Na,K-ATPase regulator which could play an important role in neuronal excitability. It has been reported that FXYD7 bears post-translationally added modifications on threonine residues and has an apparent molecular weight of 18 kDa .

Western Blot: Mouse Brain Tissue, 1:200-1:2000

Type: Primary
Antigen: FXYD7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR