Anti-COX4I2 Rabbit Polyclonal Antibody

Supplier: Proteintech
11463-1-AP-150UL
10085-058EA 509.69 USD
10085-058
Anti-COX4I2 Rabbit Polyclonal Antibody
Antibodies
COX4I2, also named as COX4L2 and COXIV-2, belongs to the cytochrome c oxidase IV family. It is one of the nuclear-coded polypeptide chains of cytochrome c oxidase, the terminal oxidase in mitochondrial electron transport. COXIV(cytochrome c oxidase IV) has two isoforms (isoform 1 and 2). Isoform 1(COX4I1) is ubiquitously expressed and isoform 2 is highly expressed in lung tissues. This antibody was generated against full length COX4I2 protein and cross reacts with COX4I1.

Western Blot: MCF7 Cells, 1:500-1:5000; IHC: Human ovary tumor Tissue, 1:20-1:200; IF: Hela Cells, 1:20-1:200; IP: HepG2 Cells, 1:500-1:5000

Type: Primary
Antigen: COX4I2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR