Anti-S100 A16 Rabbit Polyclonal Antibody

Supplier: Proteintech
11456-1-AP-150UL
10094-082EA 402 USD
10094-082
Anti-S100 A16 Rabbit Polyclonal Antibody
Antibodies
IHC: Human Colon cancer, 1:20-1:200

Type: Primary
Antigen: S100 A16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR