Anti-MZB1 Rabbit Polyclonal Antibody

Supplier: Proteintech
11454-1-AP-150UL
10090-786EA 402 USD
10090-786
Anti-MZB1 Rabbit Polyclonal Antibody
Antibodies
MZB1, also named as ,PACAP, MGC29506 and pERp1, belongs to the PERP1 family. It is associates with immunoglobulin M (IgM) heavy and light chains and promotes IgM assembly and secretion. MGC29506(PACAP) may exert its effect by acting as a molecular chaperone or as an oxidoreductase as it displays a low level of oxidoreductase activity. Isoform 2 may be involved in regulation of apoptosis. (PubMed: 11350957) MZB1 has 5 isoforms with MW 21kd and 7-13kd.

Western Blot: Raji Cells, 1:500-1:5000; IHC: Human gliomas Tissue, 1:20-1:200

Type: Primary
Antigen: MZB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR