Anti-NQO1 Rabbit Polyclonal Antibody

Supplier: Proteintech
11451-1-AP-150UL
10090-914EA 402 USD
10090-914
Anti-NQO1 Rabbit Polyclonal Antibody
Antibodies
NQO1, also named as DIA4, NMOR1, DTD and QR1, belongs to the NAD(P)H dehydrogenase (quinone) family. This enzyme apparently serves as a quinone reductase in connection with conjugation reactions of hydroquinons involved in detoxification pathways as well as in biosynthetic processes such as the vitamin K-dependent gamma-carboxylation of glutamate residues in prothrombin synthesis. It is known to be involved in benzene metabolism. In human studies of ozone exposure, polymorphisms in oxidative stress genes (NQO1, GSTM1, GSTP1) modify respiratory symptoms, lung function, biomarkers and risk of asthma. This antibody recognizes all the three isoforms (26-27 kDa and 31 kDa) of NQO1 and the homo-dimer form (66-70 kDa) of NQO1.

Western Blot:K-562 Cells, 1:500-1:5000; IHC: Human pancreas cancer Tissue, 1:20-1:200; IF: MCF-7 Cells, 1:20-1:200; IP:K-562 Cells, 1:500-1:5000; FC: MCF-7 Cells, N/A

Type: Primary
Antigen: NQO1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR