Anti-VCAM1 Rabbit Polyclonal Antibody

Supplier: Proteintech
11444-1-AP-150UL
10096-942EA 402 USD
10096-942
Anti-VCAM1 Rabbit Polyclonal Antibody
Antibodies
Vascular cell adhesion molecule 1 (VCAM1), also known as CD106, is a transmembrane glycoprotein belonging to the immunoglobulin gene superfamily. VCAM1 is expressed by cytokine-activated endothelium, interacts with integrin VLA4 (α4β1) present on the surface of leukocytes, and mediates both adhesion and signal transduction. It is also expressed either constitutively or inducibly in a variety of other cell types, including vascular smooth muscle cells, differentiating skeletal muscle cells, renal and neural epithelial cells, macrophages (Kupffer cells), dendritic cells, and bone marrow stromal cells .

IHC: Human spleen Tissue, 1:20-1:200

Type: Primary
Antigen: VCAM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR