Anti-CA9 Rabbit Polyclonal Antibody

Supplier: Proteintech
11443-1-AP-150UL
10083-926EA 402 USD
10083-926
Anti-CA9 Rabbit Polyclonal Antibody
Antibodies
CA9(Carbonic anhydrase 9) may be involved in the control of cell proliferation and transformation and appears to be a novel specific biomarker for a cervical neoplasia.It is a tumor-associated antigen that has been shown to have diagnostic utility in identifying cervical dysplasia and carcinoma.The protein is presentboth on the plasma membrane and in the nucleus of cells and has the molecular.It was observed the 54KDa and 58 Kda bands in A549(JOC,vol21,no.3,473-482).

Western Blot: Mouse Brain Tissue, 1:200-1:2000

Type: Primary
Antigen: CA9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR