Anti-COX15 Rabbit Polyclonal Antibody

Supplier: Proteintech
11441-1-AP-150UL
10085-050EA 402 USD
10085-050
Anti-COX15 Rabbit Polyclonal Antibody
Antibodies
COX15 belongs to the COX15/CtaA family. It may be involved in the biosynthesis of heme A. COX15 has 2 isoforms COX15.1 and COX15.2. The MW in SDS-PAGE is between 44kd and 55kd.

Western Blot: Mouse Skeletal Muscle Tissue, 1:500-1:5000; IHC: Human Heart, 1:20-1:200

Type: Primary
Antigen: COX15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR