Anti-COX6B2 Rabbit Polyclonal Antibody

Supplier: Proteintech
11437-1-AP-150UL
10085-070EA 402 USD
10085-070
Anti-COX6B2 Rabbit Polyclonal Antibody
Antibodies
COX6B2(Cytochrome c oxidase subunit 6B2) encodes the testis-specific cytochrome C oxidase subunit VIb. It has been shown to be expressed in a subset of lung and breast tumors at a level >1% of the testicular level of expression. It belongs to the cytochrome c oxidase subunit 6B family.

Western Blot: Mouse Heart Tissue, 1:200-1:1000; IHC: Mouse Testis Tissue, 1:20-1:200

Type: Primary
Antigen: COX6B2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR