Anti-RARG Rabbit Polyclonal Antibody

Supplier: Proteintech
11424-1-AP-150UL
10093-336EA 402 USD
10093-336
Anti-RARG Rabbit Polyclonal Antibody
Antibodies
Retinoic acid receptors (RARs) are nuclear hormone receptors that act as ligand-dependent transcriptional regulators. In their liganded state, RARs activate transcription, whereas in their nonliganded form, they repress transcription of their target genes. RARs have numerous target genes, which have retinoic response elements in their promoter regions . RARG is a member of the retinoid acid receptor (RAR) family, along with RARA, which is known to be involved PML/RARA fusion of acute promyelocytic leukemia, and RARB. and rearrangement that conferred to leukemic cells acute promyelocytic leukemia–like morphologic and immunophenotypic features .

Western Blot: HeLa Cells, 1:200-1:2000

Type: Primary
Antigen: RARG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR