Anti-COX6A2 Rabbit Polyclonal Antibody

Supplier: Proteintech
11421-1-AP-150UL
10085-066EA 402 USD
10085-066
Anti-COX6A2 Rabbit Polyclonal Antibody
Antibodies
COX6A2(Cytochrome c oxidase subunit 6A2, mitochondrial) is also named as COX6A, COX6AH and Belongs to the cytochrome c oxidase subunit 6A family. This protein is one of the nuclear-coded polypeptide chains of cytochrome c oxidase, the terminal oxidase in mitochondrial electron transport.

Type: Primary
Antigen: COX6A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR