Anti-RAB9A Rabbit Polyclonal Antibody

Supplier: Proteintech
11420-1-AP-150UL
10093-230EA 402 USD
10093-230
Anti-RAB9A Rabbit Polyclonal Antibody
Antibodies
RAB9A belongs to the small GTPase superfamily, the members of which play an important role in regulating vesicular trafficking and reside in specific intracellular compartments. RAB9A has been localized to components of the endocytic/exocytic pathway and has been implicated in recycling of membrane receptors, such as mannose 6-phosphate receptor, from early endosomes to the trans Golgi network.

Western Blot: HeLa Cells, 1:200-1:2000; IHC: Human Prostate Cancer Tissue, 1:20-1:200

Type: Primary
Antigen: RAB9A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR