Anti-COX7B Rabbit Polyclonal Antibody

Supplier: Proteintech
11417-2-AP-150UL
10085-080EA 402 USD
10085-080
Anti-COX7B Rabbit Polyclonal Antibody
Antibodies
COX7B, also named as Cytochrome c oxidase subunit 7B, mitochondrial, is a 80 amino acid protein, which belongs to the cytochrome c oxidase VIIb family. COX7B protein is one of the nuclear-coded polypeptide chains of cytochrome c oxidase, the terminal oxidase in mitochondrial electron transport. COX7B plays a role in proper central nervous system development in vertebrates. COX7B as a terminal enzyme in the mitochondrial respiratory chain (oxidative phosphorylation OXPHOS), catalyzing the electron transfer from reduced cytochrome c to molecule oxygen and plays a role in eye development.

Western Blot: Human Skeletal Muscle Tissue, 1:200-1:1000; IHC: Human Lymphoma Tissue, 1:20-1:200

Type: Primary
Antigen: COX7B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR