Anti-COX7C Rabbit Polyclonal Antibody

Supplier: Proteintech
11411-2-AP-150UL
10085-082EA 402 USD
10085-082
Anti-COX7C Rabbit Polyclonal Antibody
Antibodies
COX7C(Cytochrome c oxidase subunit 7C, mitochondrial) is the terminal enzyme in the mitochondrial respiratory chain (oxidative phosphorylation OXPHOS), catalyzing the elctron transfer from reduced cytochrome c to molecule oxygen. This protein belongs to the cytochrome c oxidase VIIc family. The full length protein has a transit peptide with 16 amino acids. This antibody is specifc to COX7C.

IHC: Human gliomas Tissue, 1:20-1:200; IF: MCF-7 Cells, 1:20-1:200; Western Blot: Mouse ovary Tissue, 1:500-1:5000

Type: Primary
Antigen: COX7C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR