Anti-EPCR Rabbit Polyclonal Antibody

Supplier: Proteintech
11391-1-AP-150UL
10086-564EA 402 USD
10086-564
Anti-EPCR Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human spleen Tissue, 1:200-1:2000; IHC: Human pancreas cancer Tissue, 1:20-1:200

Type: Primary
Antigen: EPCR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR