Anti-PSMC4 Rabbit Polyclonal Antibody

Supplier: Proteintech
11389-1-AP-150UL
10092-920EA 537.7 USD
10092-920
Anti-PSMC4 Rabbit Polyclonal Antibody
Antibodies
PSMC4(proteasome (prosome, macropain) 26S subunit, ATPase, 4) is also named as TBP7, S6, TBP7, MIP224 and belongs to the AAA ATPase family. The 26S protease is involved in the ATP-dependent degradation of ubiquitinated proteins. The predicted 458-amino acid TBP7 protein is 57% identical to TBP1 and they can form heterodimers.

Western Blot: Human Skeletal Muscle Tissue, 1:500-1:5000; IHC: Human Colon cancer Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: PSMC4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR