Anti-SULT1A4 Rabbit Polyclonal Antibody

Supplier: Proteintech
11376-1-AP-150UL
10095-382EA 402 USD
10095-382
Anti-SULT1A4 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Colon Tissue, 1:500-1:5000; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: SULT1A4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR