Anti-RYBP Rabbit Polyclonal Antibody

Supplier: Proteintech
11365-1-AP-150UL
10093-742EA 402 USD
10093-742
Anti-RYBP Rabbit Polyclonal Antibody
Antibodies
Polycomb repressive complexes (PRCs) are important chromatin regulators of embryonic stem (ES) cell function . RYBP binds Polycomb H2A monoubiquitin ligases Ring1A and Ring1B and has been suggested to assist PRC localization to their targets. RYBP, firstly identified as a direct interactor with Ring1A, acts as transcriptional repressor in reporter assays, both in tissue culture cells and in the fly

Western Blot:K-562 Cells, 1:200-1:2000

Type: Primary
Antigen: RYBP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR