Anti-MCRS1 Rabbit Polyclonal Antibody

Supplier: Proteintech
11362-1-AP-150UL
10090-060EA 402 USD
10090-060
Anti-MCRS1 Rabbit Polyclonal Antibody
Antibodies
MCRS1, also named INO80Q, MSP58 or p78, is a nucleolar protein expressed in the very early S phase. It modulates the transcription repressor activity of DAXX by recruiting it to the nucleolus. MCRS1 is also present on polyribosomes of synaptoneurosome as a novel RNA-binding protein to activate the rRNA transcription. The forkhead-associated domain of MCRS1 is involved in association with centrosomal protein Nde1, comprising a component of the centrosome and taking an essential role in viability. Its isoform MCRS2 might be a linker between telomere maintenance and cell-cycle regulation by interacting with LPTS/PinX1, a telomerase-inhibitory protein. MCRS1 is detected by Western blot as a highly modified protein at 78 kDa and minor bands at about 62 and 55 kDa.

Western Blot: HeLa Cells, 1:500-1:5000

Type: Primary
Antigen: MCRS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR