Anti-Dynamitin Rabbit Polyclonal Antibody

Supplier: Proteintech
11361-1-AP-150UL
10086-196EA 402 USD
10086-196
Anti-Dynamitin Rabbit Polyclonal Antibody
Antibodies
Dynamitin is a 50 kDa protein containing a calmodulin binding domain, a putative ATPase domain and MacMARCKS-binding domain. This protein is a part of the dynactin complex believed to link the dynactin complex to membrane compartments. Its functions are tightly associated with dynein motor protein, thus extend to vesicle trafficking and membrane integrity. Dynamitin was named so because its overexpression causes dynactin complex which contains 10 subunits, to disassemble. Its N terminal 58 amino acid is for MacMARCKS binding and residues 59-83 is responsible for calmodulin binding. This antibody is against the full length p50 dynamitin.

Western Blot: Mouse Testis Tissue, 1:200-1:2000; IHC: Human gliomas Tissue, 1:20-1:200; FC: HeLa Cells, N/A

Type: Primary
Antigen: Dynamitin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR