Anti-fibrosin Rabbit Polyclonal Antibody

Supplier: Proteintech
11356-1-AP-150UL
10087-020EA 402 USD
10087-020
Anti-fibrosin Rabbit Polyclonal Antibody
Antibodies
Fibrosin is a lymphokine secreted by activated lymphocytes that induces fibroblast proliferation FBRS (or FBS1) gene, encoding fibrosin (or fibrosin-1), was first cloned by Prakash et al. and was found to be able to stimulate several biological activities related to scarring as a fibrogenic lymphokine. Fibrosin is secreted by activated lymphocytes that induce fibroblast proliferation. In vitro application of fibrosin stimulates expression of alpha smooth muscle actin fibers and plays an important role in up regulating the appearance of myofibroblasts during wound healing and possibly in fibrotic diseases. Fibrosin is also implicated in alcohol-induced hepatic fibrosis in vivo.

Western Blot:Jurkat Cells, 1:200-1:2000; IHC: Human breast cancer Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: fibrosin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR