Anti-ADP-Ribosyltransferase 5 Rabbit Polyclonal Antibody

Supplier: Proteintech
11354-1-AP-150UL
10083-172EA 402 USD
10083-172
Anti-ADP-Ribosyltransferase 5 Rabbit Polyclonal Antibody
Antibodies
ART5 (Ecto-ADP-ribosyltransferase 5) belongs to a class of enzymes that catalyze the transfer of an ADP-ribose group from NAD to arginine residues on target proteins like G proteins. ART5 is a secreted protein. This antibody detects the 43kD ART5 protein in mouse liver and testis.

Western Blot: Mouse Testis Tissue, 1:200-1:2000; IHC: Human Heart, 1:20-1:200

Type: Primary
Antigen: [delete]
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR