Anti-WHSC1L1 Rabbit Polyclonal Antibody

Supplier: Proteintech
11345-1-AP-150UL
10091-314EA 524.34 USD
10091-314
Anti-WHSC1L1 Rabbit Polyclonal Antibody
Antibodies
WHSC1L1, also named as NSD3 and DC28, belongs to the histone-lysine methyltransferase family and SET2 subfamily. It is histone methyltransferase. WHSC1L1 is a potently transforming oncogenes based on the number of altered phenotypes expressed by the cells. It contains a PWWP-domain that is a methyl-lysine recognition motif involved in histone code modification and epigenetic regulation of gene expression. Knockdown of WHSC1L1 in 8p11-12–amplified breast cancer cells resulted in profound loss of growth and survival of these cells. . This antibody recognize the long isoform (160-170 kDa) and short isoform (68-72 kDa).

Western Blot: HeLa Cells, 1:200-1:2000; IHC: Human Lymphoma Tissue, 1:20-1:200

Type: Primary
Antigen: WHSC1L1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR